Tisk…
Protective hoods with visor for PAPR, half, FH1
Dodavatel: Scott Safety
SCTS2022948EA 2490 CZK
SCTS2022948
Protective hoods with visor for PAPR, half, FH1
Polomasky a filtrační respirátory Systémy polomasek
Ultra light weight and comfortable half hood with integrated eye protection. This FH1 has been designed with a unique neoprene face seal to deliver the highest level of PAPR protection without compromising user comfort.
  • Revolutionary soft head cradle for intuitive and straightforward donning
  • Ultra clear PETG visor giving wide field of view and a clear down-view panel
  • Half hood design with skin-friendly neoprene chin seal
Certifikace: EN 12941 TH2/3, EN 166 1F and AS/NZS1716-P2