Tisk…
Full face masks, FM3
Dodavatel: Scott Safety
SCTS2023647EA 4850 CZK
SCTS2023647
Full face masks, FM3
Halo-butyl or silicone mask for use with powered air respirator Spirit. The full facemask FM3 with a T-bar face seal design and clear inner mask provides the highest levels of comfort and wearer acceptance​.
  • Wide panoramic visor to maximise the field of vision and a front mounted speech diaphragm for easy communication
  • Side entry hose improves comfort and balance
  • Approved for unpowered use with Pro2000 filters
  • Compatible blower units: Spirit, Tornado, Proflow2 120SC, 120EX, 160SC / 160EX and 120EX Proflow3
Certifikace: EN 12942, EN 14942, EN 136
Informace pro objednávání: FM3 is supplied without hose; to be ordered separately.