Tisk…
Anti-GREM1 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9626EA 13000 CZK
BSBTPB9626
Anti-GREM1 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Gremlin-1(GREM1) detection. Tested with WB in Human;Rat.
Gremlin, also known as Drm, is a highly conserved 20.7-kDa, 184 amino acid glycoprotein part of the DAN family and is a cysteine knot-secreted protein. Skeletal cells synthesize bone morphogenetic proteins (BMPs) and BMP antagonists. And Gremlin is expressed in osteoblasts and opposes BMP effects on osteoblastic differentiation and function in vitro. Gremlin 1 (GREM 1) is known for its antagonistic reaction with BMPs in the TGF beta signaling pathway. This gene inhibits BMP-2, BMP-4, and BMP-7. Inhibition by grem 1 of BMPs in mice allow the expression of fibroblast growth factors (FGFs) 4 and 8 and Sonic hedgehog (SHH) which are necessary for proper limb development. Gremlin 1 may play an oncogenic role especially in carcinomas of the uterine cervix, lung, ovary, kidney, breast, colon, pancreas, and sarcoma. Over-expressed gremlin 1 functions by interaction with YWHAH (Its binding site for gremlin 1 was located between residues 61-80 and gremlin 1 binding site for YWHAH was found to be located between residues 1-67). Therefore, Gremlin 1 and its binding protein YWHAH could be good targets for developing diagnostic and therapeutic strategies against human cancers.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.