Tisk…
Anti-HHEX Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9631EA 16320 CZK
BSBTPB9631
Anti-HHEX Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Hematopoietically-expressed homeobox protein Hhex(HHEX) detection. Tested with WB in Human;Mouse; Rat.
Hematopoietically-expressed homeobox protein HHEX is a protein that in humans is encoded by the HHEX gene. Homeobox genes are members of a family of transcription factors that regulate tissue development in many different organisms. Hromas et al. (1993) set out to identify homeobox genes that might play a role in hematopoiesis. And using somatic cell hybrid analysis, they mapped the HHEX gene to chromosome 10, where the HOX11 gene is located. Homeobox genes are involved in neoplastic transformation of both epithelial and hemopoietic tissues. The divergent homeobox gene HEX is expressed in the anterior visceral endoderm during early mouse development and in some adult tissues of endodermal origin, including liver and thyroid. D'Elia et al.’s findings suggested that regulation of HEX entry in the nucleus of thyrocytes may represent a critical step during human thyroid tumorigenesis.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.