Tisk…
Anti-HSF4 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9634EA 16320 CZK
BSBTPB9634
Anti-HSF4 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Heat shock factor protein 4(HSF4) detection. Tested with WB in Human.
Heat shock factor protein 4 is a protein that in humans is encoded by the HSF4 gene. It is mapped to 16q21. Heat-shock transcription factors (HSFs) activate heat-shock response genes under conditions of heat or other stresses. HSF4 lacks the carboxyl-terminal hydrophobic repeat which is shared among all vertebrate HSFs and has been suggested to be involved in the negative regulation of DNA binding activity. Two alternatively spliced transcripts encoding distinct isoforms and possessing different transcriptional activity have been described.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.