Tisk…
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9636EA 15510 CZK
BSBTPB9636
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Heat shock protein HSP 90-beta(HSP90AB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Heat shock protein HSP 90-beta, also called HSP90beta, is a protein that in humans is encoded by the HSP90AB1 gene. It is mapped to chromosome 6p21.1. This gene encodes a member of the heat shock protein 90 family; these proteins are involved in signal transduction, protein folding and degradation and morphological evolution. And this gene is thought to play a role in gastric apoptosis and inflammation. Alternative splicing results in multiple transcript variants. Pseudogenes have been identified on multiple chromosomes.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.