Tisk…
Anti-HSP90B1 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9637EA 13000 CZK
BSBTPB9637
Anti-HSP90B1 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Endoplasmin(HSP90B1) detection. Tested with WB in Human;Mouse;Rat.
Heat shock protein 90kDa beta member 1 (HSP90B1), known as endoplasmin, or GRP94, is a chaperone protein that in humans is encoded by the HSP90B1 gene. It is mapped to chromosome 12q23.3. This gene encodes a member of a family of adenosine triphosphate (ATP)-metabolizing molecular chaperones with roles in stabilizing and folding other proteins. The encoded protein is localized to melanosomes and the endoplasmic reticulum. Expression of this protein is associated with a variety of pathogenic states, including tumor formation.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.