Tisk…
Anti-KCNA2 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9649EA 13000 CZK
BSBTPB9649
Anti-KCNA2 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Potassium voltage-gated channel subfamily A member 2(KCNA2) detection. Tested with WB in Human;Mouse;Rat.
Potassium voltage-gated channel subfamily A member 2, also known as Kv1.2, is a protein that in humans is encoded by the KCNA2 gene. Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. The coding region of this gene is intronless, and the gene is clustered with genes KCNA3 and KCNA10 on chromosome 1.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.