Tisk…
Anti-NLRC4 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9673EA 13000 CZK
BSBTPB9673
Anti-NLRC4 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for NLR family CARD domain-containing protein 4(NLRC4) detection. Tested with WB in Human;Mouse;Rat.
NLR family CARD domain-containing protein 4, also known as CARD12, is a protein that in humans is encoded by the NLRC4 gene. It is mapped to 2p22.3. This gene encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.