Tisk…
Anti-S100A9 Rabbit Polyclonal Antibody
Dodavatel: Boster
BSBTPB9677EA 14410 CZK
BSBTPB9677
Anti-S100A9 Rabbit Polyclonal Antibody
Protilátky
Rabbit IgG polyclonal antibody for Protein S100-A9(S100A9) detection. Tested with WB, IHC-P, ELISA in Mouse;Rat.
S100 calcium-binding protein A9 (S100A9), also known as migration inhibitory factor-related protein 14 (MRP14) or calgranulin B, is a protein that in humans is encoded by the S100A9 gene. S100-A9 is a member of the S100 family of proteins containing 2 EF hand calcium-binding motifs. And S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.