Tisk…

Anti-IgG Donkey Polyclonal Antibody (DyLight® 800)

Dodavatel: Immunoreagents
IMMRIR2890EA 7810 CZK
IMMRIR2890
Anti-IgG Donkey Polyclonal Antibody (DyLight® 800)
Protilátky
The ImmunoReagents' DyLight® 800 conjugates uses a near-IR fluorphore that is invisible to the naked eye but increases the staining options when using infrared imaging systems. DyLight® 800 has spectral properties that are very similar to other near-IR dyes, including Alexa Fluor 790 and IRDye 800, and has high solubility for maximal incorporation of the dye into the biomolecule of interest. Excitation/Emission = 777nm / 794nm Emission Color = Near IR (Similar Dyes: Alexa Fluor 790, IRDye 800)

This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR