Tisk…

Anti-Cardiac Troponin, I subunit Goat Polyclonal Antibody

Dodavatel: Immunoreagents
IMMRIR2915EA 6690 CZK
IMMRIR2915
Anti-Cardiac Troponin, I subunit Goat Polyclonal Antibody
Protilátky
  

This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications

Type: Primary
Antigen: Cardiac Troponin, I subunit
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR