Tisk…

Anti-IgG Goat Polyclonal Antibody (DyLight® 405)

Dodavatel: Immunoreagents
IMMRIR2929EA 1520 CZK
IMMRIR2929
Anti-IgG Goat Polyclonal Antibody (DyLight® 405)
Protilátky
The ImmunoReagents' DyLight 405 has a vibrant blue fluorescence intensity that replaces Alexa Fluor™ 405, Cascade™ Blue and AMCA in many applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight® 405 (Ex = 400 nm; Em = 420 nm)

This conjugate is suitable for all immunoassay applications. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:500-1:5,000 for Immunohistochemistry · 1:200-1:5000 for ELISA/Western blot

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR