Tisk…
Respiratory helmets with visors for PAPR, FH6 and FH61 to FH65
Dodavatel: Scott Safety
SCTS2023068EA 6910 CZK
SCTS2023068
Respiratory helmets with visors for PAPR, FH6 and FH61 to FH65
Polomasky a filtrační respirátory Systémy polomasek
Integrated helmet and visor with optional ear defenders. Ideal for use in hazardous substances polluted environments such as workplaces in grinding services, quarries, foundries, in construction and shipyards.
  • Choice of visor materials, coatings and shades
  • Safety helmet and rail-mounted retractable visor with flexible chin seal
  • Filter unit - filters out ambient atmosphere (with fan-air supply)
  • Compatible blower units: Spirit, Proflow2 160SC / 160EX
Certifikace: EN 12941 TH3 (PAPR), EN 397 (head protection), EN 352-3 (hearing protection), EN 166 1B9 (eye and face protection) and EN 175 (welding protection for FH61 helmet).
Informace pro objednávání: FH6 is supplied without hose; to be ordered seperately.