These synthetic surgical gloves are manufactured without the use of accelerator chemicals (thiurams mercaptobenzothiazole, diphenylthiourea, diphenylguanidine1, dithiocarbamates). Recommended for all surgical procedures or surgeries where latex allergies and/or chemical sensitivities are a concern for the patients or clinicians.
- Powder-free, micro-textured surface
- Hydrogel polymer coating
- Micro-roughened surface for improved grip
- Every glove (100%) is air inflation tested and visually inspected for quality and safety
- Non pyrogenic, potentially reducing the risk of post-operative complications
Certifikace: EN 455; AQL 0,65
Balení: One pair per pack, in a high quality inner wrap, packed into a film pack (constructed of a laminate of polyester and low-density polyethylene).