Tisk…
Surgical gloves, polychloroprene, sterile, Biogel® NeoDerm®
Dodavatel: Mölnlycke
Biogel®
MOLN42990EA 23260 CZK
MOLN42990 MOLN42980 MOLN42970 MOLN42960 MOLN42965 MOLN42985 MOLN42975
Surgical gloves, polychloroprene, sterile, Biogel® NeoDerm®
Rukavice
These synthetic surgical gloves are manufactured without the use of accelerator chemicals (thiurams mercaptobenzothiazole, diphenylthiourea, diphenylguanidine1, dithiocarbamates). Recommended for all surgical procedures or surgeries where latex allergies and/or chemical sensitivities are a concern for the patients or clinicians.
  • Powder-free, micro-textured surface
  • Hydrogel polymer coating
  • Micro-roughened surface for improved grip
  • Every glove (100%) is air inflation tested and visually inspected for quality and safety
  • Non pyrogenic, potentially reducing the risk of post-operative complications
Certifikace: EN 455; AQL 0,65
Balení: One pair per pack, in a high quality inner wrap, packed into a film pack (constructed of a laminate of polyester and low-density polyethylene).