Tisk…

Anti-RbcL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Agrisera
AGRIAS03037-HRPEA 12380 CZK
AGRIAS03037-HRP
Anti-RbcL Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Rubisco (Ribulose-1,5-bisphosphate carboxylase/oxygenase) catalyzes the rate-limiting step of CO2 fixation in photosynthetic organisms. It is demonstrably homologous from purple bacteria to flowering plants and consists of two protein subunits, each present in 8 copies. In plants and green algae, the large subunit (~55 kDa) is coded by the chloroplast rbcL gene, and the small subunit (15 kDa) is coded by a family of nuclear rbcS genes.,Rubisco (Ribulose-1,5-bisphosphate carboxylase/oxygenase) catalyzes the rate-limiting step of CO2 fixation in photosynthetic organisms. It is demonstrably homologous from purple bacteria to flowering plants and consists of two protein subunits, each present in 8 copies. In plants and green algae, the large subunit (~55 kDa) is coded by the chloroplast rbcL gene, and the small subunit (15 kDa) is coded by a family of nuclear rbcS genes.

Type: Primary
Antigen: RBCL
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Apium graveolens, A. thaliana, Artemisia annua, Baculogypsina sphaerulata, Bienertia sinuspersici, C. raudensis, C. reinhardtii, Cicer arietinum, Colobanthus quitensis kunt bartl, Cyanophora paradoxa, Cylindrospermopsis raciborskii cs-505, Dinoflagellate endosymbionts, Emiliana huxleyi, Euglena gracilis, M. fumariolicum, Fraxinus mandshurica, Fucus vesiculosus, Glycine max, Gonyaulax polyedra, Guzmania hybrid, Heterosigma akashiwo, Karenia brevis, Moestrup, Chicken, Liquidambar formosana, Micromonas pusila, N. benthamiana, Physcomitrella patens, Porphyra sp., Prochlorococcus sp., Schima superba, Spinacia oleracea, S. pinnata, Symbiodinium sp., Synechococcus, Thalassiosira pseudonana, T. punctigera, T. elongatus, T. aestivum, Vitis vinifera
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR