Tisk…

Anti-HSP90 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS05063EA 11390 CZK
AGRIAS05063
Anti-HSP90 Rabbit Polyclonal Antibody
Protilátky
HSP90 is a ubiquitous chaperone involved in numerous cellular processes. Members of this family of proteins play important roles in allowing a selected group of intracellular signaling molecules reach and maintain functionally active conformations.Alternative names: for hsp90 alpha: HSP 86, renal carcinoma antigen NY-REN-38, for hsp90 beta: hsp84,HSP90 is a ubiquitous chaperone involved in numerous cellular processes. Members of this family of proteins play important roles in allowing a selected group of intracellular signaling molecules reach and maintain functionally active conformations.Alternative names: for hsp90 alpha: HSP 86, renal carcinoma antigen NY-REN-38, for hsp90 beta: hsp84

Type: Primary
Antigen: HSP90
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Cucumis sativus, E. teft, H. vulgare, Silene vulgaris, Solanum lycopersicum, P. strobus, Zea mays, Desmodesmus subspicatus, phycobiont: Trebouxia TR1 and TR9, Plasmodium falciparum, A. thaliana, Cucumis sativus, E. teft, H. vulgare, Solanum lycopersicum, P. strobus, Zea mays, Desmodesmus subspicatus, phycobiont: Trebouxia TR1 and TR9
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR