Tisk…

Anti-Bovine;Goat;Human;Mouse;Rabbit IgM µ Goat Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS101162EA 5500 CZK
AGRIAS101162
Anti-Bovine;Goat;Human;Mouse;Rabbit IgM µ Goat Polyclonal Antibody
Protilátky
Goat anti-mouse IgM, (µ chain) affinity purified is a secondary antibody which binds to mouse IgM in immunological assay. Antibodies are affinity purified using solid phase mouse IgM.,Goat anti-mouse IgM, (µ chain) affinity purified is a secondary antibody which binds to mouse IgM in immunological assay. Antibodies are affinity purified using solid phase mouse IgM.

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Goat, Human, Mouse, Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR