Tisk…

Anti-kappa light chain Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Agrisera
AGRIAS101405EA 6210 CZK
AGRIAS101405
Anti-kappa light chain Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Goat anti-human kappa (k) chain is a secondary antibody conjugated to FITC, which binds to human kappa chain in immunological assays. Antibody is affinity purified using solid phase human kappa chain. Fluorescein-5-isothiocyanate (FITC) has Amax = 494 nm, Emax = 518 nm.,Goat anti-human kappa (k) chain is a secondary antibody conjugated to FITC, which binds to human kappa chain in immunological assays. Antibody is affinity purified using solid phase human kappa chain. Fluorescein-5-isothiocyanate (FITC) has Amax = 494 nm, Emax = 518 nm.

Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR