Tisk…

Anti-Aflatoxin M1 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS111741EA 40590 CZK
AGRIAS111741
Anti-Aflatoxin M1 Rabbit Polyclonal Antibody
Protilátky
Aflatoxins are naturally occurring mycotoxins that are produced by many species of Aspergillus. Aflatoxins are toxic and among the most carcinogenic substances known. Crops which are frequently affected by Asperiillus include cereals (maize, sorghum, pearl millet, rice, wheat), oilseeds (peanut, soybean, sunflower, cotton), spices (chile peppers, black pepper, coriander, turmeric, ginger), and tree nuts (almond, pistachio, walnut, coconut, brazil nut). ,Aflatoxins are naturally occurring mycotoxins that are produced by many species of Aspergillus. Aflatoxins are toxic and among the most carcinogenic substances known. Crops which are frequently affected by Asperiillus include cereals (maize, sorghum, pearl millet, rice, wheat), oilseeds (peanut, soybean, sunflower, cotton), spices (chile peppers, black pepper, coriander, turmeric, ginger), and tree nuts (almond, pistachio, walnut, coconut, brazil nut).

Type: Primary
Antigen: Aflatoxin M1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR