Tisk…

Anti-PSB33 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS121852EA 8070 CZK
AGRIAS121852
Anti-PSB33 Rabbit Polyclonal Antibody
Protilátky
PSB33 or TEF5 is a Rieske (2Fe-2S) domain-containing protein located in chloroplast thylakoid membrane. The protein has oxireductase activity and is involved in oxidation reaction. ,PSB33 or TEF5 is a Rieske (2Fe-2S) domain-containing protein located in chloroplast thylakoid membrane. The protein has oxireductase activity and is involved in oxidation reaction.

Type: Primary
Antigen: PSB33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Spinacia oleracea
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR