Tisk…

Anti-FLS2 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS121857EA 8070 CZK
AGRIAS121857
Anti-FLS2 Rabbit Polyclonal Antibody
Protilátky
FLS2 (Flagellin-sensitive 2) is a pattern-recognition receptor and a single-pass membrane protein. Alternative names: FLAGELLIN-SENSITIVE 2, FLAGELLIN-SENSING 2, FLS2, LRR receptor-like serine/threonine-protein kinase FLS2,FLS2 (Flagellin-sensitive 2) is a pattern-recognition receptor and a single-pass membrane protein. Alternative names: FLAGELLIN-SENSITIVE 2, FLAGELLIN-SENSING 2, FLS2, LRR receptor-like serine/threonine-protein kinase FLS2

Type: Primary
Antigen: FLS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Ficus glabrata
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR