Tisk…

Anti-EIN2 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS121865EA 8070 CZK
AGRIAS121865
Anti-EIN2 Rabbit Polyclonal Antibody
Protilátky
EIN2 (Ethylene-insensitive protein 2) is a central factor in signaling pathways regulated by ethylene including various processes like: plant defense and development, senescence, nucleotide sugar flux and tropisms. Alternative names: EIN2, ETHYLENE INSENSITIVE 2, PIR2, CKR1, CYTOKININ RESISTANT 1, ERA3, ENHANCED RESPONSE TO ABA3, ORE3, ORESARA 3, ORE2, ORESARA 2, ATEIN2.,EIN2 (Ethylene-insensitive protein 2) is a central factor in signaling pathways regulated by ethylene including various processes like: plant defense and development, senescence, nucleotide sugar flux and tropisms. Alternative names: EIN2, ETHYLENE INSENSITIVE 2, PIR2, CKR1, CYTOKININ RESISTANT 1, ERA3, ENHANCED RESPONSE TO ABA3, ORE3, ORESARA 3, ORE2, ORESARA 2, ATEIN2.

Type: Primary
Antigen: EIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Synechocystis sp. pcc 6803
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR