Tisk…

Anti-NRT2.1 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS122612EA 9000 CZK
AGRIAS122612
Anti-NRT2.1 Rabbit Polyclonal Antibody
Protilátky
NRT2.1 is a high affinity nitrate transporter which is up-regulated by nitrate. Functions as a repressor of lateral root initiation independently of nitrate uptake. Synonymes: Protein ACH1, Protein LATERAL ROOT INITIATION 1, ACH1, ATNRT2.1, ATNRT2:1,, LIN1, NITRATE TRANSPORTER 2, NITRATE TRANSPORTER 2.1, NITRATE TRANSPORTER 2:1, NRT2, NRT2.1, NRT2:1, NRT2;1AT,NRT2.1 is a high affinity nitrate transporter which is up-regulated by nitrate. Functions as a repressor of lateral root initiation independently of nitrate uptake. Synonymes: Protein ACH1, Protein LATERAL ROOT INITIATION 1, ACH1, ATNRT2.1, ATNRT2:1,, LIN1, NITRATE TRANSPORTER 2, NITRATE TRANSPORTER 2.1, NITRATE TRANSPORTER 2:1, NRT2, NRT2.1, NRT2:1, NRT2;1AT

Type: Primary
Antigen: NRT2.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR