Tisk…

Anti-VPS29 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132633EA 8070 CZK
AGRIAS132633
Anti-VPS29 Rabbit Polyclonal Antibody
Protilátky
VPS29 (vacuolar protein sorting 29) is a part of a retromer-like protein complex involved in endosome to lysosome protein transport. Synonymes:Vacuolar protein sorting-associated protein 29, Protein MAIGO 1.,VPS29 (vacuolar protein sorting 29) is a part of a retromer-like protein complex involved in endosome to lysosome protein transport. Synonymes:Vacuolar protein sorting-associated protein 29, Protein MAIGO 1.

Type: Primary
Antigen: VPS29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR