Tisk…

Anti-PYR1 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132634EA 8070 CZK
AGRIAS132634
Anti-PYR1 Rabbit Polyclonal Antibody
Protilátky
PYR1 (Abscisic acid receptor RCAR11) is a member of PYR (pyrabactin resistance) family, which function as abscisic acid sensors. Alternative names: ABI1-binding protein 6, Protein PYRABACTIN RESISTANCE 1, Regulatory components of ABA receptor 11.,PYR1 (Abscisic acid receptor RCAR11) is a member of PYR (pyrabactin resistance) family, which function as abscisic acid sensors. Alternative names: ABI1-binding protein 6, Protein PYRABACTIN RESISTANCE 1, Regulatory components of ABA receptor 11.

Type: Primary
Antigen: PYR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR