Tisk…

Anti-ZTL Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132662EA 8070 CZK
AGRIAS132662
Anti-ZTL Rabbit Polyclonal Antibody
Protilátky
ZTL Zeitlupe (Adagio Protein 1) is a component of an E3 ubiquitin ligase complex which is playing a central role in blue light-dependent circadian cycles. Alternative names: Adagio protein 1 (ADO1), F-box only protein 2b (FBX2b), Flavin-binding kelch repeat F-box protein 1-like protein 2, FKF1-like protein 2 (FKL2), LOV kelch protein 1 (LKP1), Clock-associated PAS protein ZTL, ZEITLUPE (ZTL)., ZTL Zeitlupe (Adagio Protein 1) is a component of an E3 ubiquitin ligase complex which is playing a central role in blue light-dependent circadian cycles. Alternative names: Adagio protein 1 (ADO1), F-box only protein 2b (FBX2b), Flavin-binding kelch repeat F-box protein 1-like protein 2, FKF1-like protein 2 (FKL2), LOV kelch protein 1 (LKP1), Clock-associated PAS protein ZTL, ZEITLUPE (ZTL).

Type: Primary
Antigen: ZTL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR