Tisk…

Anti-calcineurin/EF-hand motif Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132666EA 9000 CZK
AGRIAS132666
Anti-calcineurin/EF-hand motif Rabbit Polyclonal Antibody
Protilátky
Calcineurine is a calcium-binding protein involved in signal transduction. ,Calcineurine is a calcium-binding protein involved in signal transduction.

Type: Primary
Antigen: calcineurin, EF-hand motif
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, Populus sp., T. aestivum
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR