Tisk…

Anti-NAD(P)H dehydrogenase subunit 5 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132711EA 6290 CZK
AGRIAS132711
Anti-NAD(P)H dehydrogenase subunit 5 Rabbit Polyclonal Antibody
Protilátky
NDH (NAD(P)H-quinone oxidoreductase subunit 5, chloroplastic) is involved in the transfer of electrons from NAD(P)H:plastoquinone, via FMN and iron-sulfur centers, to quinones in the photosynthetic chain and possibly in a chloroplast respiratory chain. Alternative names: NAD(P)H-quinone oxidoreductase subunit 5, chloroplastic, NADH-plastoquinone oxidoreductase subunit 5. ,NDH (NAD(P)H-quinone oxidoreductase subunit 5, chloroplastic) is involved in the transfer of electrons from NAD(P)H:plastoquinone, via FMN and iron-sulfur centers, to quinones in the photosynthetic chain and possibly in a chloroplast respiratory chain. Alternative names: NAD(P)H-quinone oxidoreductase subunit 5, chloroplastic, NADH-plastoquinone oxidoreductase subunit 5.

Type: Primary
Antigen: NAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Oryza sativa
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR