Tisk…

Anti-GAPC2 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132727EA 9190 CZK
AGRIAS132727
Anti-GAPC2 Rabbit Polyclonal Antibody
Protilátky
Glyceraldehyde-3-phosphate dehydrogenase is an enzyme that catalyzes the first step in glycolysis by converting D-glyceraldehyde 3-phosphate (G3P) into 3-phospho-D-glyceroyl phosphate. This enzyme is essential for the carbohydrate metabolism and the maintenance of cellular ATP levels. ,Glyceraldehyde-3-phosphate dehydrogenase is an enzyme that catalyzes the first step in glycolysis by converting D-glyceraldehyde 3-phosphate (G3P) into 3-phospho-D-glyceroyl phosphate. This enzyme is essential for the carbohydrate metabolism and the maintenance of cellular ATP levels.

Type: Primary
Antigen: GAPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, N. tabacum, Solanum lycopersicum, Zea mays
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR