Tisk…

Anti-CDPK (1,2,3,4,7,8,10,11,14,20,25,30,32) Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS132754EA 8070 CZK
AGRIAS132754
Anti-CDPK (1,2,3,4,7,8,10,11,14,20,25,30,32) Rabbit Polyclonal Antibody
Protilátky
CDPK (calcium-dependent protein kinase) may play a role in signal transfuction pathways.,CDPK (calcium-dependent protein kinase) may play a role in signal transfuction pathways.

Type: Primary
Antigen: CDPK (1,2,3,4,7,8,10,11,14,20,25,30,32)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR