Tisk…

Anti-LHCSR3 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS142766EA 8070 CZK
AGRIAS142766
Anti-LHCSR3 Rabbit Polyclonal Antibody
Protilátky
LHCSR3 (Stress-related chlorophyll a/b binding protein 3) plays a role in an efficient enery dissipation process, called non-photochemical quenching (NPQ), in Chlamydomonas reinhardtii. ,LHCSR3 (Stress-related chlorophyll a/b binding protein 3) plays a role in an efficient enery dissipation process, called non-photochemical quenching (NPQ), in Chlamydomonas reinhardtii.

Type: Primary
Antigen: LHCSR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR