Tisk…

Anti-ZCP1 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS142770EA 8070 CZK
AGRIAS142770
Anti-ZCP1 Rabbit Polyclonal Antibody
Protilátky
ZCP1 belongs to a large family of putative metallochaperones, likned with maintainance of zinc homeostasis in all living organisms.,ZCP1 belongs to a large family of putative metallochaperones, likned with maintainance of zinc homeostasis in all living organisms.

Type: Primary
Antigen: ZCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR