Tisk…

Anti-GOX Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS142772EA 8070 CZK
AGRIAS142772
Anti-GOX Rabbit Polyclonal Antibody
Protilátky
Glycolate oxidase (GOX) is an enzyme which modulates reactive oxygen species-mediated signal transduction during nonhost resistance. There are three GOX isoforms in Arabidopsis thaliana: GOX1,2,3. Alternative name: Peroxisomal (S)-2-hydroxy-acid oxidase GLO.,Glycolate oxidase (GOX) is an enzyme which modulates reactive oxygen species-mediated signal transduction during nonhost resistance. There are three GOX isoforms in Arabidopsis thaliana: GOX1,2,3. Alternative name: Peroxisomal (S)-2-hydroxy-acid oxidase GLO.

Type: Primary
Antigen: GOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, Miscanthus giganteus, Phaseolus vulgaris, Spartina sp., Zea mays
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR