Tisk…

Anti-Aflatoxin M1 Mouse Monoclonal Antibody

Dodavatel: Agrisera
AGRIAS142801-3MGEA 126900 CZK
AGRIAS142801-3MG
Anti-Aflatoxin M1 Mouse Monoclonal Antibody
Protilátky
Aflatoxins are naturally occurring mycotoxins that are produced by many species of Aspergillus. Aflatoxins are toxic and among the most carcinogenic substances known. Crops which are frequently affected by Asperiillus include cereals (maize, sorghum, pearl millet, rice, wheat), oilseeds (peanut, soybean, sunflower, cotton), spices (chile peppers, black pepper, coriander, turmeric, ginger), and tree nuts (almond, pistachio, walnut, coconut, brazil nut).,Aflatoxins are naturally occurring mycotoxins that are produced by many species of Aspergillus. Aflatoxins are toxic and among the most carcinogenic substances known. Crops which are frequently affected by Asperiillus include cereals (maize, sorghum, pearl millet, rice, wheat), oilseeds (peanut, soybean, sunflower, cotton), spices (chile peppers, black pepper, coriander, turmeric, ginger), and tree nuts (almond, pistachio, walnut, coconut, brazil nut).

Type: Primary
Antigen: Aflatoxin M1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR