Tisk…

Anti-NBR1 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS142805AEA 9000 CZK
AGRIAS142805A
Anti-NBR1 Rabbit Polyclonal Antibody
Protilátky
NBR1 (Autophagy substrate NBR1) is involved in selective autophagy process of damaged organelles, intracellular microbes, protein aggregates, cellular structures and specific soluble proteins. NBR1 mediates the process as an autophagic adapter. The protein has two UBA domains but only the C-terminal UBA domain bound ubiquitin. Alternative names: At4g24690, Putative uncharacterized protein F22K18.110.,NBR1 (Autophagy substrate NBR1) is involved in selective autophagy process of damaged organelles, intracellular microbes, protein aggregates, cellular structures and specific soluble proteins. NBR1 mediates the process as an autophagic adapter. The protein has two UBA domains but only the C-terminal UBA domain bound ubiquitin. Alternative names: At4g24690, Putative uncharacterized protein F22K18.110.

Type: Primary
Antigen: NBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR