Tisk…

Anti-MIP1 Rabbit Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS152826EA 8070 CZK
AGRIAS152826
Anti-MIP1 Rabbit Polyclonal Antibody
Protilátky
MIP1 (Aquaporin, glycerol transport activity) is localized to the contractile vacuole, which in most freshwater flagellates is used to expel excess water.,MIP1 (Aquaporin, glycerol transport activity) is localized to the contractile vacuole, which in most freshwater flagellates is used to expel excess water.
Reacts with Chlamydomonas reinhardtii (strains CC3395, and UVM4 from Neupert et al., 2009. (GeneRation of Chlamydomonas Strains that Efficiently Express Nuclear Transgenes.)

Type: Primary
Antigen: MIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR