Tisk…

Anti-IgG (H+L) Llama Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Agrisera
AGRIAS152886EA 4940 CZK
AGRIAS152886
Anti-IgG (H+L) Llama Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Llama anti-Mouse IgG (H&L), HRP Conjugated is a secondary antibody which binds to heavy (?) chains chains on mouse IgG light chains on all mouse immunoglobulins ,Llama anti-Mouse IgG (H&L), HRP Conjugated is a secondary antibody which binds to heavy (?) chains chains on mouse IgG light chains on all mouse immunoglobulins

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR