Tisk…

Anti-IgG (H+L) Goat Polyclonal Antibody

Dodavatel: Agrisera
AGRIAS152901EA 5500 CZK
AGRIAS152901
Anti-IgG (H+L) Goat Polyclonal Antibody
Protilátky
Goat anti-dog IgG is a secondary antibody which binds to all dog IgGs in immunological assays.,Goat anti-dog IgG is a secondary antibody which binds to all dog IgGs in immunological assays.

Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR