Tisk…

Anti-6x His Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: AD1.1.10]

Dodavatel: Agrisera
AGRIAS152935EA 11480 CZK
AGRIAS152935
Anti-6x His Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: AD1.1.10]
Protilátky
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine and is directly conjugated to ALP, Alkaline Phosphatase. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. ,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine and is directly conjugated to ALP, Alkaline Phosphatase. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana, Sauromatum guttatum, T. aestivum
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR