Tisk…

Anti-6x His Mouse Monoclonal Antibody [clone: AD1.1.10]

Dodavatel: Agrisera
AGRIAS152936EA 19740 CZK
AGRIAS152936
Anti-6x His Mouse Monoclonal Antibody [clone: AD1.1.10]
Protilátky
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is not conjugated to any dye or enzyme. ,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is not conjugated to any dye or enzyme.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR