Tisk…
Protective hoods, with visor, Flowhood FH51
Dodavatel: Scott Safety
SCTS2023116EA 10450 CZK
SCTS2023116
Protective hoods, with visor, Flowhood FH51
The Flowhood 51 (FH51) is a full hood with helmet and integral visor and has been developed to offer integrated protection in higher risk environments including demolition, toxic and hazardous materials handling and environmental cleanup.
  • Respirator, hard hat, and face protector all in one system
  • Hypalon material hood with taped seams
  • Polycarbonate or acetate visor options
  • Molten metal splash resistant
  • Coverage for head and shoulders, integrates with protective clothing
Applications may be found in the chemical, construction, nuclear and pharmaceutical industries. Additionally, in conjunction with multi hazard filters the Flowhood 51 would find application in extended duration response work after major incidents.
Certifikace: EN 12941 TH3, EN 397 (head protection) and EN 166 1B9 (eye and face protection).
Informace pro objednávání: FH51 is supplied without hose and that should be ordered separately.