Tisk…
Temperature loggers, CurveX-2 USB oven logger
SIMECX1500EA 69690 CZK
SIMECX1500
Temperature loggers, CurveX-2 USB oven logger
CurveX-2 USB oven datalogger offers temperature logging for industrial paint and powdercoat cure ovens. It includes ideal finish analysis software to analyse the logged temperature data and create detailed reports. Ideal for industrial oven and laboratory oven temperature profiling such as cure data analysis, ideal cure and tolerance bands.
  • Easy to use
  • Large and quick display of measurement results
Informace o dodávce: Supplied with software, insulation box (300 °C), energy absorber, silicone gasket, probe identification kit and carrying case.