Tisk…
Face shield, Interchange™ Browguard IB2000
Dodavatel: Scott Safety
Interchange™
SCTS2019716EA 0 CZK
SCTS2019716
Face shield, Interchange™ Browguard IB2000
Štíty obličejové
Interchange™ Browguard IB2000 is designed with a durable nylon shell construction to provide a high protection and comfort level in industrial applications where eye and face protection are required. Its uniquely designed flexible hammer-head clips make the fitting and removal of visors both quick and intuitive.
  • Compatible with extensive range of Interchange visors
  • Easy to adjust ratchet headgear with comfort pad
Certifikace: EN 166; EN 1731S
Informace pro objednávání: Visors must be ordered separately.