Tisk…

Anti-WDR27 Mouse Monoclonal Antibody [clone: 3C5]

Dodavatel: Abnova
ABNOH00253769-M01EA 11230 CZK
ABNOH00253769-M01
Anti-WDR27 Mouse Monoclonal Antibody [clone: 3C5]
Protilátky
Mouse monoclonal antibody raised against a full-length recombinant WDR27.

Type: Primary
Antigen: WDR27
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR