Tisk…

Anti-EID3 Mouse Monoclonal Antibody [clone: 2E5]

Dodavatel: Abnova
ABNOH00493861-M08EA 10890 CZK
ABNOH00493861-M08
Anti-EID3 Mouse Monoclonal Antibody [clone: 2E5]
Protilátky
Mouse monoclonal antibody raised against a full-length recombinant EID3.

Type: Primary
Antigen: EID3
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR