Tisk…

Anti-PTK2B Mouse Monoclonal Antibody [clone: 3G2]

Dodavatel: Abnova
ABNOH00002185-M02AEA 10890 CZK
ABNOH00002185-M02A
Anti-PTK2B Mouse Monoclonal Antibody [clone: 3G2]
Protilátky
Mouse monoclonal antibody raised against a partial recombinant PTK2B.

Type: Primary
Antigen: PTK2B
Clonality: Monoclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgA/IgG/IgM Mix lambda
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR