Tisk…

Anti-ANGPTL3 Mouse Polyclonal Antibody

Dodavatel: Abnova
ABNOH00027329-B01PEA 10890 CZK
ABNOH00027329-B01P
Anti-ANGPTL3 Mouse Polyclonal Antibody
Protilátky
Mouse polyclonal antibody raised against a full-length human ANGPTL3 protein.

Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR