Tisk…

Anti-CD3E Hamster Monoclonal Antibody [clone: 145-2C11] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Dodavatel: Abnova
ABNOMAB12205EA 10550 CZK
ABNOMAB12205
Anti-CD3E Hamster Monoclonal Antibody [clone: 145-2C11] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Protilátky
Armenian Hamster monoclonal antibody raised against Cd3e.

Type: Secondary
Antigen: Cd3e
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: PerCP/Cy5.5
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR