Tisk…

Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (FITC (Fluorescein Isothiocyanate))

Dodavatel: Abnova
ABNOMAB12209EA 10550 CZK
ABNOMAB12209
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-5] (FITC (Fluorescein Isothiocyanate))
Protilátky
Rat monoclonal antibody raised against Cd4.

Type: Secondary
Antigen: Cd4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR